missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate, tech.

CAS: 375-02-0 | C4HF7O | 198.04 g/mol

$2492.62 - $2492.62

Chemical Identifiers

CAS 375-02-0
Molecular Formula C4HF7O
Molecular Weight (g/mol) 198.04
MDL Number MFCD00039731
InChI Key IQJZGNJYXIIMGP-UHFFFAOYSA-N
Synonym heptafluorobutyraldehyde, butanal, heptafluoro, butyraldehyde, heptafluoro, heptafluorobutyraldehyde, tech., perfluorobutanal, perfluorobutyraldehyde, c3f7cho, heptafluoro-n-butyraldehyde, butanal,2,2,3,3,4,4,4-heptafluoro
PubChem CID 67809
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutanal
SMILES FC(F)(F)C(F)(F)C(F)(F)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0087514
View Documents
Thermo Scientific Chemicals
L0087514
25 g
Each for $2,492.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

375-02-0
198.04
IQJZGNJYXIIMGP-UHFFFAOYSA-N
67809
FC(F)(F)C(F)(F)C(F)(F)C=O
C4HF7O
MFCD00039731
heptafluorobutyraldehyde, butanal, heptafluoro, butyraldehyde, heptafluoro, heptafluorobutyraldehyde, tech., perfluorobutanal, perfluorobutyraldehyde, c3f7cho, heptafluoro-n-butyraldehyde, butanal,2,2,3,3,4,4,4-heptafluoro
2,2,3,3,4,4,4-heptafluorobutanal

Specifications

375-02-0
1.32
25 g
heptafluorobutyraldehyde, butanal, heptafluoro, butyraldehyde, heptafluoro, heptafluorobutyraldehyde, tech., perfluorobutanal, perfluorobutyraldehyde, c3f7cho, heptafluoro-n-butyraldehyde, butanal,2,2,3,3,4,4,4-heptafluoro
FC(F)(F)C(F)(F)C(F)(F)C=O
198.04
198.04 (Anhydrous)
2,2,3,3,4,4,4-Heptafluorobutyraldehyde hydrate
C4HF7O
MFCD00039731
1367365
IQJZGNJYXIIMGP-UHFFFAOYSA-N
2,2,3,3,4,4,4-heptafluorobutanal
67809
Technical
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 206-783-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only