missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluorobutylamine, 97%, Thermo Scientific Chemicals

Catalog No. p-7046911
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1677606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1677606 Supplier Thermo Scientific Chemicals Supplier No. L1677606
Only null left

2,2,3,3,4,4,4-Heptafluorobutylamine is used as pharmaceutical intermediates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,4-Heptafluorobutylamine is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 374-99-2
Molecular Formula C4H4F7N
Molecular Weight (g/mol) 199.072
MDL Number MFCD00014817
InChI Key WBGBQSRNXPVFDB-UHFFFAOYSA-N
Synonym 2,2,3,3,4,4,4-heptafluorobutylamine, 1h,1h-heptafluorobutylamine, heptafluorobutylamine, butylamine, 2,2,3,3,4,4,4-heptafluoro, 1-butanamine, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-perfluorobutylamine, acmc-1af94, 1-butanamine,2,2,3,3,4,4,4-heptafluoro
PubChem CID 67807
IUPAC Name 2,2,3,3,4,4,4-heptafluorobutan-1-amine
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)N

Specifications

Density 1.493
Boiling Point 70°C to 71°C
Refractive Index 1.304
Quantity 5 g
UN Number UN2735
Beilstein 1761906
Solubility Information Soluble in water.
Formula Weight 199.07
Percent Purity 97%
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluorobutylamine
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 206-780-0
RTECSNumber EO5190000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only