missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,4,4,4-Heptafluoro-1-iodobutane, 96%

Catalog No. p-7044035
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0417303 1 g
AAL0417306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0417303 Supplier Thermo Scientific Chemicals Supplier No. L0417303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 374-98-1 | C4H2F7I | 309.953 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 374-98-1
Molecular Formula C4H2F7I
Molecular Weight (g/mol) 309.953
MDL Number MFCD00013719
InChI Key AFHPVXVEXINDFS-UHFFFAOYSA-N
Synonym heptafluorobutyl iodide, 2,2,3,3,4,4,4-heptafluoro-1-iodobutane, butane, 1,1,1,2,2,3,3-heptafluoro-4-iodo, 1h,1h-heptafluorobutyl iodide, 1h,1h-heptafluoro-1-iodobutane, acmc-20aoko, 1-iodo-1h,1h-perfluorobutane, 1h,1h-heptafluoro-1-iodobutan, 1-iodo-2,2,3,3,4,4,4-heptafluorobutane
PubChem CID 67806
IUPAC Name 1,1,1,2,2,3,3-heptafluoro-4-iodobutane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)I

Specifications

Density 2.054
Boiling Point 91°C to 92°C
Flash Point None
Refractive Index 1.3625
Linear Formula CF3CF2CF2CH2I
Quantity 1 g
Beilstein 1761927
Sensitivity Light sensitive
Formula Weight 309.95
Percent Purity 96%
Chemical Name or Material 2,2,3,3,4,4,4-Heptafluoro-1-iodobutane
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-779-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only