missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoropropylamine 97.0+%, TCI America™

Catalog No. P22815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22815G 5 g
P22811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22815G Supplier TCI America Supplier No. P22815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 422-03-7
Molecular Formula C3H4F5N
Molecular Weight (g/mol) 149.064
MDL Number MFCD00042459
InChI Key DPQNQLKPUVWGHE-UHFFFAOYSA-N
PubChem CID 79002
IUPAC Name 2,2,3,3,3-pentafluoropropan-1-amine
SMILES C(C(C(F)(F)F)(F)F)N

Specifications

Color Colorless
Boiling Point 50°C
Quantity 5 g
UN Number 2735
Synonym 2,2,3,3,3-pentafluoropropylamine, pentafluoropropylamine, acmc-1amc1, 1h,1h-pentafluoropropylamine, 2,2,3,3,3-penta-fluoropropylamine, 2,2,3,3,3-pentafluoro-propylamine, 2,2,3,3,3-pentafluoro-propyl amine, 1-propanamine,2,2,3,3,3-pentafluoro, 1-propanamine, 2,2,3,3,3-pentafluoro
Formula Weight 149.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,3-Pentafluoropropylamine