missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,3,3,3-Pentafluoropropyl Methacrylate (stabilized with TBC) 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 46°C |
| Quantity | 5 g |
| UN Number | 3272 |
| Formula Weight | 218.12 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2,3,3,3-Pentafluoropropyl Methacrylate (stabilized with TBC) |
Safety and Handling
| EINECSNumber | (2)-1031 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |