missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoropropyl acrylate, 97%

Catalog No. p-7045344
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1034303 1 g
AAL1034306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1034303 Supplier Thermo Scientific Chemicals Supplier No. L1034303

CAS: 356-86-5 | C6H5F5O2 | 204.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 356-86-5
Molecular Formula C6H5F5O2
Molecular Weight (g/mol) 204.10
MDL Number MFCD00039257
InChI Key JDVGNKIUXZQTFD-UHFFFAOYSA-N
Synonym 2,2,3,3,3-pentafluoropropyl acrylate, 1h,1h-pentafluoropropyl acrylate, 2-propenoic acid, 2,2,3,3,3-pentafluoropropyl ester, acmc-20aokp, acrylic acid=2,2,3,3,3-pentafluoropropyl ester, acrylic acid 2,2,3,3,3-pentafluoropropyl ester, 2-propenoic acid,2,2,3,3,3-pentafluoropropyl ester, 2,2,3,3,3-pentafluoropropyl acrylate stabilized with tbc, 2,2,3,3,3-pentafluoropropyl acrylate, contains 100 ppm 4-tert-butylcatechol as inhibitor
PubChem CID 67744
IUPAC Name 2,2,3,3,3-pentafluoropropyl prop-2-enoate
SMILES FC(F)(F)C(F)(F)COC(=O)C=C

Specifications

Density 1.32
Boiling Point 49°C to 50°C (100 mmHg)
Flash Point 23°C (73°F)
Odor Pungent
Refractive Index 1.336
Linear Formula H2C=CHCO2CH2CF2CF3
Quantity 1 g
UN Number UN3272
Beilstein 1779140
Formula Weight 204.1
Percent Purity 97%
Chemical Name or Material 2,2,3,3,3-Pentafluoropropyl acrylate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: II
EINECSNumber 206-608-4
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only