missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoro-1-propanol 98.0+%, TCI America™

Catalog No. P084525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P084525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P084525G Supplier TCI America Supplier No. P084525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 422-05-9
Molecular Formula C3H3F5O
Molecular Weight (g/mol) 150.048
MDL Number MFCD00004673
InChI Key PSQZJKGXDGNDFP-UHFFFAOYSA-N
Synonym 2,2,3,3,3-pentafluoro-1-propanol, pentafluoro-1-propanol, 1,1-dihydroperfluoropropanol, pfpoh, 2,2,3,3,3-pentafluoropropanol, 1h,1h-pentafluoropropanol, 1-propanol, 2,2,3,3,3-pentafluoro, 1h,1h-pentafluoropropanol-1, c2f5ch2oh, unii-epj1s961sn
PubChem CID 9872
IUPAC Name 2,2,3,3,3-pentafluoropropan-1-ol
SMILES C(C(C(F)(F)F)(F)F)O

Specifications

Color Colorless
Boiling Point 81°C
Quantity 25 g
Formula Weight 150.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,3,3,3-Pentafluoro-1-propanol
EINECSNumber (2)-3364
RTECSNumber UB8800000
TSCA Yes