missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoro-1-propanol, 97%

Catalog No. p-178665
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303790050 5 g Glass bottle
AC303790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303790050 Supplier Thermo Scientific Chemicals Supplier No. 303790050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 422-05-9 | C3H3F5O | 150.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 422-05-9
Molecular Formula C3H3F5O
Molecular Weight (g/mol) 150.05
MDL Number MFCD00004673
InChI Key PSQZJKGXDGNDFP-UHFFFAOYSA-N
Synonym 2,2,3,3,3-pentafluoro-1-propanol, pentafluoro-1-propanol, 1,1-dihydroperfluoropropanol, pfpoh, 2,2,3,3,3-pentafluoropropanol, 1h,1h-pentafluoropropanol, 1-propanol, 2,2,3,3,3-pentafluoro, 1h,1h-pentafluoropropanol-1, c2f5ch2oh, unii-epj1s961sn
PubChem CID 9872
IUPAC Name 2,2,3,3,3-pentafluoropropan-1-ol
SMILES C(C(C(F)(F)F)(F)F)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Yellow
Boiling Point 81.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.2880 to 1.2950
Linear Formula CF3CF2CH2OH
Quantity 5 g
Beilstein 01, IV, 1438
Formula Weight 150.05
Percent Purity 97%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2,2,3,3,3-Pentafluoro-1-propanol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-012-7
RTECSNumber UB8800000
TSCA TSCA

RUO – Research Use Only