missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trimethylacetamidine hydrochloride, 98%

Catalog No. AAA1061306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1061306 5 g
AAA1061314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1061306 Supplier Thermo Scientific Chemicals Supplier No. A1061306

CAS: 18202-73-8 | C5H13ClN2 | 136.623 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18202-73-8
Molecular Formula C5H13ClN2
Molecular Weight (g/mol) 136.623
MDL Number MFCD00051988
InChI Key ARDGQYVTLGUJII-UHFFFAOYSA-N
Synonym 2,2-dimethylpropanimidamide hydrochloride, pivalamidine hydrochloride, pivalimidamide hydrochloride, tert-butylcarbamidine hydrochloride, 2,2-dimethylpropionamidine hydrochloride, 2,2-dimethyl-propionamidine hydrochloride, 2,2-dimethylpropionamidine hcl, t-butylcarbamidine hydrochloride, 2,2-dimethylpropanamidine hydrochloride, 2,2,2-trimethylacetamidine hydrochloride
PubChem CID 2781880
IUPAC Name 2,2-dimethylpropanimidamide;hydrochloride
SMILES CC(C)(C)C(=N)N.Cl

Specifications

Melting Point 188°C to 193°C
Quantity 5 g
Beilstein 3618865
Sensitivity Hygroscopic
Formula Weight 136.63
Percent Purity 98%
Chemical Name or Material 2,2,2-Trimethylacetamidine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-091-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only