missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylamine 97.0+%, TCI America™

Catalog No. p-7136639
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11695G 5 g
T116925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11695G Supplier TCI America Supplier No. T11695G
Only null left

Chemical Identifiers

CAS 753-90-2
Molecular Formula C2H4F3N
Molecular Weight (g/mol) 99.056
MDL Number MFCD00008132
InChI Key KIPSRYDSZQRPEA-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethylamine, trifluoroethylamine, ethanamine, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-amine, 2-amino-1,1,1-trifluoroethane, ethylamine, 2,2,2-trifluoro, 2,2,2-trifluoro-ethylamine, 2,2,2-trifluorethanamin, trifluoro ethylamine, trifluoroethyl amine
PubChem CID 9773
IUPAC Name 2,2,2-trifluoroethanamine
SMILES C(C(F)(F)F)N

Specifications

Color Yellow
Boiling Point 37°C
Quantity 5 g
UN Number 2733
Formula Weight 99.06
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethylamine
RTECSNumber KS0175000
TSCA Yes
Recommended Storage Refrigerator