missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylamine Hydrochloride 98.0+%, TCI America™

Catalog No. T11705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11705G 5 g
T117025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T11705G Supplier TCI America Supplier No. T11705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 373-88-6
Molecular Formula C2H5ClF3N
Molecular Weight (g/mol) 135.51
MDL Number MFCD00012875
InChI Key ZTUJDPKOHPKRMO-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethylamine hydrochloride, 2,2,2-trifluoroethanamine hydrochloride, trifluoroethylamine hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride, 2,2,2-trifluoroethylaminehydrochloride, 2,2,2-trifluoroethylamine, hydrochloride, 2,2,2-trifluoroethan-1-amine hydrochloride, 2,2,2-trifluoroethylamine hcl, ethylamine, trifluoro-, hydrochloride, ethanamine, 2,2,2-trifluoro-, hydrochloride 1:1
PubChem CID 9772
IUPAC Name hydrogen 2,2,2-trifluoroethan-1-amine chloride
SMILES [H+].[Cl-].NCC(F)(F)F

Specifications

Quantity 5 g
Formula Weight 135.51
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2,2-Trifluoroethylamine Hydrochloride
RTECSNumber KS0250000
TSCA Yes