missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylamine, 99.5%

Catalog No. AC303500100
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303500100 10 g Glass bottle
AC303500010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303500100 Supplier Thermo Scientific Chemicals Supplier No. 303500100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 753-90-2 | C2H4F3N | 99.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 753-90-2
Molecular Formula C2H4F3N
Molecular Weight (g/mol) 99.06
MDL Number MFCD00008132
InChI Key KIPSRYDSZQRPEA-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethylamine, trifluoroethylamine, ethanamine, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-amine, 2-amino-1,1,1-trifluoroethane, ethylamine, 2,2,2-trifluoro, 2,2,2-trifluoro-ethylamine, 2,2,2-trifluorethanamin, trifluoro ethylamine, trifluoroethyl amine
PubChem CID 9773
IUPAC Name 2,2,2-trifluoroethanamine
SMILES C(C(F)(F)F)N

Specifications

CAS Min % 99.4
CAS Max % 100.0
Color Colorless
Density 1.2400g/mL
Boiling Point 37°C to 38°C
Flash Point -16°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Refractive Index 1.3000 to 1.302
Linear Formula CF3CH2NH2
Quantity 10 g
Specific Gravity 1.24
Formula Weight 99.06
Percent Purity 99.50%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f
EINECSNumber 212-041-3

RUO – Research Use Only