missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylamine, 98%

Catalog No. AAB2078903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2078903 1 g
AAB2078906 5 g
AAB2078914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2078903 Supplier Thermo Scientific Chemicals Supplier No. B2078903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 753-90-2 | C2H4F3N | 99.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 753-90-2
Molecular Formula C2H4F3N
Molecular Weight (g/mol) 99.056
MDL Number MFCD00008132
InChI Key KIPSRYDSZQRPEA-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethylamine, trifluoroethylamine, ethanamine, 2,2,2-trifluoro, 2,2,2-trifluoroethan-1-amine, 2-amino-1,1,1-trifluoroethane, ethylamine, 2,2,2-trifluoro, 2,2,2-trifluoro-ethylamine, 2,2,2-trifluorethanamin, trifluoro ethylamine, trifluoroethyl amine
PubChem CID 9773
IUPAC Name 2,2,2-trifluoroethanamine
SMILES C(C(F)(F)F)N

Specifications

Density 1.245
Boiling Point 36°C to 38°C
Flash Point −16°C (3°F)
Odor Amine-like
Refractive Index 1.295
Quantity 1 g
UN Number UN2733
Beilstein 1733204
Formula Weight 99.06
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethylamine
Show More Show Less
Hazard Category H225-H314-H331-H335
Hazard Statement GHS H Statement
H225-H314-H318-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 212-041-3
RTECSNumber KS0175000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only