missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl trifluoromethanesulfonate, 95%

Catalog No. AAH6160314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6160314 25 g
AAH6160306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6160314 Supplier Thermo Scientific Chemicals Supplier No. H6160314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6226-25-1 | C3H2F6O3S | 232.096 g/mol

2,2,2-Trifluoroethyl trifluoromethanesulfonate is a powerful trifluoroethylating agent which is useful for synthesis of fluorinated amino acids. It is used as a reagent in thenantioselective preparation of cyclic N-aryl hydroxamic acids via phase-transfer catalyzed alkylation of nitrobenzyl bromides to give nirophenylalanines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroethyl trifluoromethanesulfonate is a powerful trifluoroethylating agent which is useful for synthesis of fluorinated amino acids. It is used as a reagent in thenantioselective preparation of cyclic N-aryl hydroxamic acids via phase-transfer catalyzed alkylation of nitrobenzyl bromides to give nirophenylalanines.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 6226-25-1
Molecular Formula C3H2F6O3S
Molecular Weight (g/mol) 232.096
MDL Number MFCD00671579
InChI Key RTMMSCJWQYWMNK-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyltrifluoromethanesulfonate, 2,2,2-trifluoroethyl triflate, methanesulfonic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoromethanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluorometanesulfonic acid, 2,2,2-trifluoroethyltrifluoromethanesulphonate, 2,2,2-trifluoroethyl trifluoromethane sulfonate, trifluoroethyltriflate, trifluoroethyl triflate
PubChem CID 80366
IUPAC Name 2,2,2-trifluoroethyl trifluoromethanesulfonate
SMILES C(C(F)(F)F)OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Yellow
Density 1.611 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.306
Linear Formula CF3SO3CH2CF3
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 232.1
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl trifluoromethanesulfonate
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
RTECSNumber PB2775000
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only