Learn More
2,2,2-Trifluoroethyl trichloromethanesulfonate, 94%
CAS: 23199-56-6 | C3H2Cl3F3O3S | 281.45 g/mol
$241.85 - $986.81
Chemical Identifiers
| CAS | 23199-56-6 |
|---|---|
| Molecular Formula | C3H2Cl3F3O3S |
| Molecular Weight (g/mol) | 281.45 |
| MDL Number | MFCD00042400 |
| InChI Key | GOIWZZQXWJVDOG-UHFFFAOYSA-N |
| Synonym | 2,2,2-trifluoroethyl trichloromethanesulphonate, 2,2,2-trifluoroethyl trichloromethane-sulfonate, trichloromethanesulfonic acid 2,2,2-trifluoroethyl ester, methanesulfonic acid, trichloro-, 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trichloromethyl sulfonate, acmc-20aoku, trifluoroethyl trichloromethylsulfonate, trifluoroethyl trichloromethanesulphonate, 2,2,2-trifluoroethyltrichloromethanesulfonate, 1,1,1-trifluoroethyl trichloromethanesulfonate |
| PubChem CID | 90027 |
| IUPAC Name | 2,2,2-trifluoroethyl trichloromethanesulfonate |
| SMILES | FC(F)(F)COS(=O)(=O)C(Cl)(Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0863506
|
Thermo Scientific Chemicals
L0863506 |
5 g |
Each for $241.85
|
|
|||||
|
AAL0863514
|
Thermo Scientific Chemicals
L0863514 |
25 g |
Each for $986.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 23199-56-6 | |
| 281.45 | |
| GOIWZZQXWJVDOG-UHFFFAOYSA-N | |
| 90027 | |
| FC(F)(F)COS(=O)(=O)C(Cl)(Cl)Cl |
| C3H2Cl3F3O3S | |
| MFCD00042400 | |
| 2,2,2-trifluoroethyl trichloromethanesulphonate, 2,2,2-trifluoroethyl trichloromethane-sulfonate, trichloromethanesulfonic acid 2,2,2-trifluoroethyl ester, methanesulfonic acid, trichloro-, 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trichloromethyl sulfonate, acmc-20aoku, trifluoroethyl trichloromethylsulfonate, trifluoroethyl trichloromethanesulphonate, 2,2,2-trifluoroethyltrichloromethanesulfonate, 1,1,1-trifluoroethyl trichloromethanesulfonate | |
| 2,2,2-trifluoroethyl trichloromethanesulfonate |
Specifications
| 23199-56-6 | |
| 1.714 | |
| Acrid | |
| 1.422 | |
| 5 g | |
| 1108366 | |
| 2,2,2-trifluoroethyl trichloromethanesulphonate, 2,2,2-trifluoroethyl trichloromethane-sulfonate, trichloromethanesulfonic acid 2,2,2-trifluoroethyl ester, methanesulfonic acid, trichloro-, 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl trichloromethyl sulfonate, acmc-20aoku, trifluoroethyl trichloromethylsulfonate, trifluoroethyl trichloromethanesulphonate, 2,2,2-trifluoroethyltrichloromethanesulfonate, 1,1,1-trifluoroethyl trichloromethanesulfonate | |
| FC(F)(F)COS(=O)(=O)C(Cl)(Cl)Cl | |
| 281.45 | |
| 281.47 | |
| 2,2,2-Trifluoroethyl trichloromethanesulfonate |
| -70°C | |
| 84°C to 86°C (20 mmHg) | |
| C3H2Cl3F3O3S | |
| MFCD00042400 | |
| UN3265 | |
| Moisture sensitive | |
| GOIWZZQXWJVDOG-UHFFFAOYSA-N | |
| 2,2,2-trifluoroethyl trichloromethanesulfonate | |
| 90027 | |
| 94% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 245-484-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only