missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoroethyl Nonafluorobutanesulfonate 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 141°C |
| Quantity | 5 g |
| Formula Weight | 382.12 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,2,2-Trifluoroethyl Nonafluorobutanesulfonate |
Safety and Handling
| TSCA | No |