missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoroethyl Methanesulfonate 98.0+%, TCI America™
$29.83 - $72.70
Chemical Identifiers
| CAS | 25236-64-0 |
|---|---|
| Molecular Formula | C3H5F3O3S |
| Molecular Weight (g/mol) | 178.13 |
| MDL Number | MFCD00025058 |
| InChI Key | ICECLJDLAVVEOW-UHFFFAOYSA-N |
| Synonym | 2,2,2-trifluoromethyl mesylate, methanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl methanesulphonate, 2,2,2-trifluoroethyl mesylate, 2,2,2-trifluoroethyl methylsulfonate, acmc-209gib, cf3ch2oso2ch3, icecljdlavveow-uhfffaoysa, 2,2,2-trifluoroethyl methansulfonate, 2,2,2 trifluoroethyl methanesulfonate |
| PubChem CID | 556565 |
| IUPAC Name | 2,2,2-trifluoroethyl methanesulfonate |
| SMILES | CS(=O)(=O)OCC(F)(F)F |
Chemical Identifiers
| 25236-64-0 | |
| 178.13 | |
| ICECLJDLAVVEOW-UHFFFAOYSA-N | |
| 556565 | |
| CS(=O)(=O)OCC(F)(F)F |
| C3H5F3O3S | |
| MFCD00025058 | |
| 2,2,2-trifluoromethyl mesylate, methanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl methanesulphonate, 2,2,2-trifluoroethyl mesylate, 2,2,2-trifluoroethyl methylsulfonate, acmc-209gib, cf3ch2oso2ch3, icecljdlavveow-uhfffaoysa, 2,2,2-trifluoroethyl methansulfonate, 2,2,2 trifluoroethyl methanesulfonate | |
| 2,2,2-trifluoroethyl methanesulfonate |
Specifications
| 25236-64-0 | |
| 99°C | |
| MFCD00025058 | |
| 3265 | |
| ICECLJDLAVVEOW-UHFFFAOYSA-N | |
| 2,2,2-trifluoroethyl methanesulfonate | |
| 556565 | |
| ≥98.0% (GC) | |
| 2,2,2-Trifluoroethyl Methanesulfonate |
| Colorless | |
| C3H5F3O3S | |
| 5 g | |
| 2,2,2-trifluoromethyl mesylate, methanesulfonic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyl methanesulphonate, 2,2,2-trifluoroethyl mesylate, 2,2,2-trifluoroethyl methylsulfonate, acmc-209gib, cf3ch2oso2ch3, icecljdlavveow-uhfffaoysa, 2,2,2-trifluoroethyl methansulfonate, 2,2,2 trifluoroethyl methanesulfonate | |
| CS(=O)(=O)OCC(F)(F)F | |
| 178.13 | |
| 178.13 | |
| Liquid |
Safety and Handling
TSCA : No