missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl Formate 95.0+%, TCI America™

Catalog No. p-7137298
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25861G 1 g
T25865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25861G Supplier TCI America Supplier No. T25861G

Chemical Identifiers

CAS 32042-38-9
Molecular Formula C3H3F3O2
Molecular Weight (g/mol) 128.05
MDL Number MFCD06797733
InChI Key CAFROQYMUICGNO-UHFFFAOYSA-N
Synonym tfef, formic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylformate, trifluoroethyl formate, zlchem 326, hcooch2cf3, acmc-209hq8, ksc495g7p, 2,2,2-trifluoroethyl formate, 2,2,2-tris fluoranyl ethyl methanoate
PubChem CID 141681
IUPAC Name 2,2,2-trifluoroethyl formate
SMILES FC(F)(F)COC=O

Specifications

Color Yellow
Boiling Point 60°C
Quantity 1 g
UN Number 3272
Formula Weight 128.05
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl Formate