missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl formate, 95%, Thermo Scientific Chemicals

Catalog No. AAH2721203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2721203 1 g
AAH2721206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2721203 Supplier Thermo Scientific Chemicals Supplier No. H2721203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32042-38-9
Molecular Formula C3H3F3O2
Molecular Weight (g/mol) 128.05
MDL Number MFCD06797733
InChI Key CAFROQYMUICGNO-UHFFFAOYSA-N
Synonym tfef, formic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethylformate, trifluoroethyl formate, zlchem 326, hcooch2cf3, acmc-209hq8, ksc495g7p, 2,2,2-trifluoroethyl formate, 2,2,2-tris fluoranyl ethyl methanoate
PubChem CID 141681
IUPAC Name 2,2,2-trifluoroethyl formate
SMILES FC(F)(F)COC=O

Specifications

Density 1.317
Boiling Point 59°C to 60°C
Flash Point 7°C (45°F)
Refractive Index 1.001
Quantity 1 g
UN Number UN3272
Formula Weight 128.05
Percent Purity 95%
Chemical Name or Material 2,2,2-Trifluoroethyl formate
Hazard Category H225-H302+H332-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only