missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. p-7117270
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11525G 5 g
A115225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11525G Supplier TCI America Supplier No. A11525G
Only null left

Chemical Identifiers

CAS 407-47-6
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.088
MDL Number MFCD00000444
InChI Key VBHXIMACZBQHPX-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl acrylate, 2-propenoic acid, 2,2,2-trifluoroethyl ester, 2-propenoic acid, trifluoroethyl ester, 2,2,2-trifluoroethylacrylate, acrylic acid, 2,2,2-trifluoroethyl ester, acrylic acid 2,2,2-trifluoroethyl ester, tfol-a, acmc-20aokw, trifluoroethyl acrylate, pubchem12647
PubChem CID 67889
IUPAC Name 2,2,2-trifluoroethyl prop-2-enoate
SMILES C=CC(=O)OCC(F)(F)F

Specifications

Color Colorless
Boiling Point 92°C
Quantity 5 g
UN Number 3272
Formula Weight 154.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl Acrylate (stabilized with MEHQ)