missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl acrylate, 98%, stab. with 200ppm 4-methoxyphenol

Catalog No. p-7024060
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1823506 5 g
AAA1823514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1823506 Supplier Thermo Scientific Chemicals Supplier No. A1823506
Only null left

CAS: 407-47-6 | C5H5F3O2 | 154.088 g/mol

It is used for adhesives because of its peeling property and also used for contact lens to give antifouling property. It is also employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for adhesives because of its peeling property and also used for contact lens to give antifouling property. It is also employed as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents, heat, light UV radiation and free radical initiators. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from light and electrostatic charges. Keep it refrigerated.

Chemical Identifiers

CAS 407-47-6
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.088
MDL Number MFCD00000444
InChI Key VBHXIMACZBQHPX-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl acrylate, 2-propenoic acid, 2,2,2-trifluoroethyl ester, 2-propenoic acid, trifluoroethyl ester, 2,2,2-trifluoroethylacrylate, acrylic acid, 2,2,2-trifluoroethyl ester, acrylic acid 2,2,2-trifluoroethyl ester, tfol-a, acmc-20aokw, trifluoroethyl acrylate, pubchem12647
PubChem CID 67889
IUPAC Name 2,2,2-trifluoroethyl prop-2-enoate
SMILES C=CC(=O)OCC(F)(F)F

Specifications

Density 1.216
Boiling Point 91°C to 93°C
Flash Point 12°C (53°F)
Odor Pungent
Refractive Index 1.35
Linear Formula CF3CH2O2CCH=CH2
Quantity 5 g
UN Number UN3272
Beilstein 1756611
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 154.09
Percent Purity 98%
Chemical Name or Material 2,2,2-Trifluoroethyl acrylate, stabilized with 200ppm 4-methoxyphenol
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 206-987-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only