missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethyl Acetate 97.0+%, TCI America™

Catalog No. T34225G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T34225G 5 g
T342225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T34225G Supplier TCI America Supplier No. T34225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 406-95-1
Molecular Formula C4H5F3O2
Molecular Weight (g/mol) 142.08
MDL Number MFCD00198018
InChI Key ZOWSJJBOQDKOHI-UHFFFAOYSA-N
Synonym trifluoroethyl acetate, unii-en07rvl78u, en07rvl78u, acetic acid, 2,2,2-trifluoroethyl ester, acetic acid 2,2,2-trifluoroethyl ester, trifluoroethylacetate, 2,2,2-trifluoroethylacetate, acmc-20aokx, cf3ch2o2cch3, ksc494s2j
PubChem CID 136256
IUPAC Name 2,2,2-trifluoroethyl acetate
SMILES CC(=O)OCC(F)(F)F

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
UN Number 3272
Formula Weight 142.08
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoroethyl Acetate