missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoro-N,N-dimethylacetamide 98.0+%, TCI America™
Supplier: TCI America T326225G
Specifications
| 2,2,2-Trifluoro-N,N-dimethylacetamide | |
| Colorless | |
| C4H6F3NO | |
| 25 g | |
| WXBWKMLIVXELSF-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-N,N-dimethylacetamide | |
| 350491 | |
| ≥98.0% (GC) |
| 1547-87-1 | |
| 136°C | |
| MFCD00043555 | |
| 1993 | |
| CN(C)C(=O)C(F)(F)F | |
| 141.09 | |
| 141.09 | |
| Liquid |
Chemical Identifiers
| 1547-87-1 | |
| 141.09 | |
| WXBWKMLIVXELSF-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-N,N-dimethylacetamide |
| C4H6F3NO | |
| MFCD00043555 | |
| 350491 | |
| CN(C)C(=O)C(F)(F)F |
Safety and Handling
TSCA : No