missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoro-4'-methylacetophenone 97.0+%, TCI America™

Catalog No. T19731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19731G 1 g
T19735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19731G Supplier TCI America Supplier No. T19731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 394-59-2
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00052338
InChI Key DYILUJUELMWXAL-UHFFFAOYSA-N
Synonym 4-trifluoroacetyl toluene, 2,2,2-trifluoro-1-p-tolyl ethanone, 4'-methyl-2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-p-tolylethanone, 2,2,2-trifluoro-1-4-methylphenyl ethanone, 2,2,2-trifluoro-4'-methylacetophenone, 2,2,2-trifluoro-1-4-methylphenyl ethan-1-one, 2,2,2-trifluoro-1-p-tolyl-ethanone, 4-trifluoroacetyltoluene, acmc-209j5m
PubChem CID 136237
IUPAC Name 2,2,2-trifluoro-1-(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 1 g
Formula Weight 188.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,2-Trifluoro-4′-methylacetophenone