missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trifluoro-4'-methoxyacetophenone 98.0+%, TCI America™
Supplier: TCI America T19865G
Specifications
| 2,2,2-Trifluoro-4′-methoxyacetophenone | |
| Yellow | |
| MFCD00018061 | |
| 4′C-Methoxy-2,2,2-trifluoroacetophenone, 4-(Trifluoroacetyl)anisole | |
| COC1=CC=C(C=C1)C(=O)C(F)(F)F | |
| 204.148 | |
| 204.15 | |
| Liquid |
| 711-38-6 | |
| C9H7F3O2 | |
| 5 g | |
| NCJZVRPXSSYDBG-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-(4-methoxyphenyl)ethanone | |
| 136555 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 711-38-6 | |
| 204.148 | |
| NCJZVRPXSSYDBG-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-(4-methoxyphenyl)ethanone |
| C9H7F3O2 | |
| MFCD00018061 | |
| 136555 | |
| COC1=CC=C(C=C1)C(=O)C(F)(F)F |
Safety and Handling
TSCA : No