missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride, 95%, Thermo Scientific Chemicals

Catalog No. p-7031588
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3271703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3271703 Supplier Thermo Scientific Chemicals Supplier No. H3271703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1138011-22-9 | Molecular Formula: C7H8ClF3N2 | Molecular Weight: 212.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1138011-22-9
Molecular Formula C7H8ClF3N2
Molecular Weight (g/mol) 212.6
MDL Number MFCD06739079
InChI Key NLDVAGWIUPSGQC-UHFFFAOYSA-N
Synonym 2,2,2-trifluoro-1-pyridin-3-yl ethanamine hydrochloride, +/--2,2,2-trifluoro-1-pyridin-3-ylethylamine hydrochloride, 2,2,2-trifluoro-1-pyridin-3-ylethylamine hydrochloride, 2,2,2-trifluoro-1-pyridin-3-yl-ethylamine hydrochloride, 2,2,2-trifluoro-1-3-pyridyl ethylamine hydrochloride, acmc-20aokz, 2,2,2-trifluoro-1-3-pyridinyl ethanamine hydrochloride, 2,2,2-trifluoro-1-3-pyridinyl ethylamine hydrochloride, 2,2,2-tris fluoranyl-1-pyridin-3-yl-ethanamine hydrochloride
PubChem CID 45594310
IUPAC Name 2,2,2-trifluoro-1-pyridin-3-ylethanamine;hydrochloride
SMILES C1=CC(=CN=C1)C(C(F)(F)F)N.Cl

Specifications

Melting Point 97°C to 102°C
Quantity 1 g
Formula Weight 212.6
Percent Purity 95%
Chemical Name or Material 2,2,2-Trifluoro-1-(3-pyridyl)ethylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only