missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Tribromoethanol 98.0+%, TCI America™
$121.28 - $121.28
Chemical Identifiers
| CAS | 75-80-9 |
|---|---|
| Molecular Formula | C2H3Br3O |
| Molecular Weight (g/mol) | 282.757 |
| MDL Number | MFCD00004671 |
| InChI Key | YFDSDPIBEUFTMI-UHFFFAOYSA-N |
| Synonym | tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan |
| PubChem CID | 6400 |
| IUPAC Name | 2,2,2-tribromoethanol |
| SMILES | C(C(Br)(Br)Br)O |
Chemical Identifiers
| 75-80-9 | |
| 282.757 | |
| YFDSDPIBEUFTMI-UHFFFAOYSA-N | |
| 6400 | |
| C(C(Br)(Br)Br)O |
| C2H3Br3O | |
| MFCD00004671 | |
| tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan | |
| 2,2,2-tribromoethanol |
Specifications
| 75-80-9 | |
| White-Yellow | |
| C2H3Br3O | |
| 25 g | |
| YFDSDPIBEUFTMI-UHFFFAOYSA-N | |
| 2,2,2-tribromoethanol | |
| 6400 | |
| ≥98.0% (GC) | |
| 2,2,2-Tribromoethanol |
| 81°C | |
| 94°C | |
| MFCD00004671 | |
| tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan | |
| C(C(Br)(Br)Br)O | |
| 282.757 | |
| 282.76 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-2002
RTECSNumber : KM3675000
TSCA : Yes