missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Tribromoethanol, 99%

CAS: 75-80-9 | C2H3Br3O | 282.76 g/mol

Supplier:  Thermo Scientific Chemicals 421430500

 View more versions of this product

Catalog No. AC421430500

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 2, 2-Tribromoethanol
98.5
73°C to 79°C
Glass bottle
Br3CCH2OH
50 g
11,111
tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
YFDSDPIBEUFTMI-UHFFFAOYSA-N
2,2,2-tribromoethanol
6400
99%
75-80-9
100.0
98.5% min. (GC)
C2H3Br3O
MFCD00004671
01, II, 338
12, 9743
Solubility in water: 25g/L @ 40°C
C(C(Br)(Br)Br)O
282.76
282.76

Chemical Identifiers

75-80-9
282.76
YFDSDPIBEUFTMI-UHFFFAOYSA-N
6400
C(C(Br)(Br)Br)O
C2H3Br3O
MFCD00004671
tribromoethanol, avertin, bromethol, ethobrome, tribromethanol, ethobrom, basibrol, narcolan, narcotyl, narkolan
2,2,2-tribromoethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-903-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only