missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(2-Propynyloxy)ethoxy]ethylamine 98.0+%, TCI America™

Catalog No. P22251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22251G 1 g
P22255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22251G Supplier TCI America Supplier No. P22251G
Only null left

Chemical Identifiers

CAS 944561-44-8
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
InChI Key GVHLQHHQXWSDDM-UHFFFAOYSA-N
Synonym 2-[2-(Propargyloxy)ethoxy]ethylamine
PubChem CID 55219291
IUPAC Name 2-(2-prop-2-ynoxyethoxy)ethanamine
SMILES C#CCOCCOCCN

Specifications

Color Yellow
Quantity 1 g
UN Number 2735
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-[2-(2-Propynyloxy)ethoxy]ethylamine
TSCA No
Recommended Storage Refrigerator