missing translation for 'onlineSavingsMsg'
Learn More

2-(2,2-Diethoxyethyl)-1,3-propanediol 95.0+%, TCI America™

Catalog No. D24375G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24375G Supplier TCI America Supplier No. D24375G
Only null left

Chemical Identifiers

CAS 55387-85-4
Molecular Formula C9H20O4
Molecular Weight (g/mol) 192.255
MDL Number MFCD00671524
InChI Key HKZYWLNVGYRMDG-UHFFFAOYSA-N
Synonym 3,3-Bis(hydroxymethyl)propionaldehyde Diethyl Acetal, 3,3-Diethoxy-1,1-bis(hydroxymethyl)propane, 4,4-Diethoxy-2-hydroxymethylbutanol
PubChem CID 12200648
IUPAC Name 2-(2,2-diethoxyethyl)propane-1,3-diol
SMILES CCOC(CC(CO)CO)OCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 192.26
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2,2-Diethoxyethyl)-1,3-propanediol