missing translation for 'onlineSavingsMsg'
Learn More

2-{2-[(2-Bromo-6-chlorophenyl)amino]phenyl}acetic Acid, MilliporeSigma™ Supelco™

Catalog No. 111022363
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022363 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022363 Supplier Merck Emd Millipore Supplier No. 0045650MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127792-23-8
Molecular Formula C14H11BrClNO2
Molecular Weight (g/mol) 340.60
MDL Number MFCD19704935
InChI Key RABKUCPRNJPBRH-UHFFFAOYSA-N
Synonym Diclofenac impurity D (PhEur); 2-[(2-Bromo-6-chlorophenyl)amino]benzeneacetic acid
IUPAC Name 2-{2-[(2-bromo-6-chlorophenyl)amino]phenyl}acetic acid
SMILES OC(=O)CC1=CC=CC=C1NC1=C(Cl)C=CC=C1Br

Specifications

Linear Formula C14H11BrClNO2
UN Number UN2811 - Class 6.1 - PG 3 - EHS - Toxic solids, organic, n.o.s., HI: all
Quantity 50 mg
Formula Weight 340.60
Percent Purity ≥98% (HPLC)
Hazard Category H301 - H315 - H319 - H411
ShelfLife Limited shelf life, expiry date on the label