missing translation for 'onlineSavingsMsg'
Learn More

2,2,2,4'-Tetrafluoroacetophenone 98.0+%, TCI America™

Catalog No. T32575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32575G 5 g
T325725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32575G Supplier TCI America Supplier No. T32575G
Only null left

Chemical Identifiers

CAS 655-32-3
Molecular Formula C8H4F4O
Molecular Weight (g/mol) 192.11
MDL Number MFCD00042393
InChI Key LUKLMXJAEKXROG-UHFFFAOYSA-N
PubChem CID 69564
IUPAC Name 2,2,2-trifluoro-1-(4-fluorophenyl)ethan-1-one
SMILES FC1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

Color White-Yellow
Boiling Point 152°C
Quantity 5 g
UN Number 1325
Formula Weight 192.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,2,2,4′-Tetrafluoroacetophenone
TSCA Yes
Recommended Storage Refrigerator