Learn More
2,2,2,3'-Tetrafluoroacetophenone, 98%
CAS: 708-64-5 | C8H4F4O | 192.11 g/mol
$56.66 - $227.73
Chemical Identifiers
| CAS | 708-64-5 |
|---|---|
| Molecular Formula | C8H4F4O |
| Molecular Weight (g/mol) | 192.11 |
| MDL Number | MFCD00039236 |
| InChI Key | NQSPTMFCJGKOQJ-UHFFFAOYSA-N |
| Synonym | 2,2,2,3'-tetrafluoroacetophenone, 2,2,2-trifluoro-1-3-fluorophenyl ethanone, m-fluorotrifluoroacetophenone, acetophenone, 2,2,2,3'-tetrafluoro, 2,2,2-trifluoro-1-3-fluorophenyl ethan-1-one, ethanone, 2,2,2-trifluoro-1-3-fluorophenyl, ethanone,2,2,2-trifluoro-1-3-fluorophenyl, acmc-20aol4, m,alpha,alpha,alpha-tetrafluoroacetophenone, 2,2,2,3'-trtrafluoroacetophenone |
| PubChem CID | 69723 |
| IUPAC Name | 2,2,2-trifluoro-1-(3-fluorophenyl)ethanone |
| SMILES | FC1=CC=CC(=C1)C(=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6470403
|
Thermo Scientific Chemicals
H6470403 |
1 g |
Each for $56.66
|
|
|||||
|
AAH6470406
|
Thermo Scientific Chemicals
H6470406 |
5 g |
Each for $227.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 708-64-5 | |
| 192.11 | |
| NQSPTMFCJGKOQJ-UHFFFAOYSA-N | |
| 69723 | |
| FC1=CC=CC(=C1)C(=O)C(F)(F)F |
| C8H4F4O | |
| MFCD00039236 | |
| 2,2,2,3'-tetrafluoroacetophenone, 2,2,2-trifluoro-1-3-fluorophenyl ethanone, m-fluorotrifluoroacetophenone, acetophenone, 2,2,2,3'-tetrafluoro, 2,2,2-trifluoro-1-3-fluorophenyl ethan-1-one, ethanone, 2,2,2-trifluoro-1-3-fluorophenyl, ethanone,2,2,2-trifluoro-1-3-fluorophenyl, acmc-20aol4, m,alpha,alpha,alpha-tetrafluoroacetophenone, 2,2,2,3'-trtrafluoroacetophenone | |
| 2,2,2-trifluoro-1-(3-fluorophenyl)ethanone |
Specifications
| 708-64-5 | |
| 1.372 g/mL | |
| 17°C (63°F) | |
| 1.444 | |
| 1 g | |
| 1872788 | |
| NQSPTMFCJGKOQJ-UHFFFAOYSA-N | |
| 2,2,2-trifluoro-1-(3-fluorophenyl)ethanone | |
| 69723 | |
| 98% | |
| 2,2,2,3'-Tetrafluoroacetophenone |
| Colorless to Yellow | |
| 79°C (50 mmHg) | |
| C8H4F4O | |
| MFCD00039236 | |
| UN1224 | |
| 2,2,2,3'-tetrafluoroacetophenone, 2,2,2-trifluoro-1-3-fluorophenyl ethanone, m-fluorotrifluoroacetophenone, acetophenone, 2,2,2,3'-tetrafluoro, 2,2,2-trifluoro-1-3-fluorophenyl ethan-1-one, ethanone, 2,2,2-trifluoro-1-3-fluorophenyl, ethanone,2,2,2-trifluoro-1-3-fluorophenyl, acmc-20aol4, m,alpha,alpha,alpha-tetrafluoroacetophenone, 2,2,2,3'-trtrafluoroacetophenone | |
| FC1=CC=CC(=C1)C(=O)C(F)(F)F | |
| 192.11 | |
| 192.11 | |
| Liquid |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 211-903-6
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only