missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl Methanesulfonate 98.0+%, TCI America™

Catalog No. T35055G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35055G 5 g
T35051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35055G Supplier TCI America Supplier No. T35055G

Chemical Identifiers

CAS 160969-03-9
Molecular Formula C11H13F3O5S
Molecular Weight (g/mol) 314.275
MDL Number MFCD16038209
InChI Key HOJMCBMXHWZNKX-UHFFFAOYSA-N
Synonym Methanesulfonic Acid 2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl Ester
PubChem CID 10519307
IUPAC Name 2-[2-(2,2,2-trifluoroethoxy)phenoxy]ethyl methanesulfonate
SMILES CS(=O)(=O)OCCOC1=CC=CC=C1OCC(F)(F)F

Specifications

Melting Point 54°C
Color White
Quantity 5 g
Formula Weight 314.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[2-(2,2,2-Trifluoroethoxy)phenoxy]ethyl Methanesulfonate