missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Pyrazol-3-yl)pyridine 98.0+%, TCI America™

Catalog No. P22111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22111G 1 g
P22115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22111G Supplier TCI America Supplier No. P22111G

Chemical Identifiers

CAS 75415-03-1
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD00115151
InChI Key HKEWOTUTAYJWQJ-UHFFFAOYSA-N
Synonym 2-1h-pyrazol-3-yl pyridine, 2-1h-pyrazol-5-yl pyridine, 2-pyrazol-3-yl-pyridine, 2-1h-pyrazol-3-yl-pyridine, 2-3-pyrazolyl pyridine, pyridine, 2-1h-pyrazol-3-yl, 3-2-pyridyl pyrazole, 2-2h-pyrazol-3-yl pyridine, 2-3e-pyrazol-3-ylidene-1h-pyridine, pyrazolylpyridine
PubChem CID 2797657
IUPAC Name 2-(1H-pyrazol-5-yl)pyridine
SMILES N1N=CC=C1C1=CC=CC=N1

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
Formula Weight 145.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1H-Pyrazol-3-yl)pyridine