missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Pyrazol-3-yl)pyridine, 98%

Catalog No. p-7033539
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5067003 1 g
AAH5067006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5067003 Supplier Thermo Scientific Chemicals Supplier No. H5067003
Only null left

CAS: 75415-03-1 | C8H7N3 | 145.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 75415-03-1
Molecular Formula C8H7N3
Molecular Weight (g/mol) 145.17
MDL Number MFCD00115151
InChI Key HKEWOTUTAYJWQJ-UHFFFAOYSA-N
Synonym 2-1h-pyrazol-3-yl pyridine, 2-1h-pyrazol-5-yl pyridine, 2-pyrazol-3-yl-pyridine, 2-1h-pyrazol-3-yl-pyridine, 2-3-pyrazolyl pyridine, pyridine, 2-1h-pyrazol-3-yl, 3-2-pyridyl pyrazole, 2-2h-pyrazol-3-yl pyridine, 2-3e-pyrazol-3-ylidene-1h-pyridine, pyrazolylpyridine
PubChem CID 2797657
IUPAC Name 2-(1H-pyrazol-5-yl)pyridine
SMILES N1N=CC=C1C1=CC=CC=N1

Specifications

Melting Point 121°C to 122°C
Quantity 1 g
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material 2-(1H-Pyrazol-3-yl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only