missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-Pyrazol-1-yl)benzoic acid, ≥97%, Thermo Scientific™

Catalog No. p-3110134
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC24401CB 250mg
CC24401DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC24401CB Supplier Thermo Scientific Chemicals Supplier No. CC24401CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55317-53-8
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD03086184
InChI Key MHACZVWKWUMHRR-UHFFFAOYSA-N
Synonym 2-1h-pyrazol-1-yl benzoic acid, 2-1-pyrazolyl benzoic acid, 2-pyrazol-1-yl benzoic acid, benzoic acid, 2-1h-pyrazol-1-yl, 2-pyrazol-1-yl-benzoic acid, 2-pyrazolylbenzoic acid, pubchem19561, 2-1-pyrazolyl benzoicacid, benzoic acid,2-1h-pyrazol-1-yl
PubChem CID 4738383
IUPAC Name 2-pyrazol-1-ylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)N2C=CC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250mg
Formula Weight 188.19
Percent Purity 97%
Chemical Name or Material 2-(1H-pyrazol-1-yl)benzoic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.