missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-triazol-1-ylmethyl)aniline, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC47514CB

 View more versions of this product

Catalog No. CC47514CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-(1H-1, 2, 4-Triazol-1-ylmethyl)aniline
96.5
97%
C9H10N4
250mg
CBGQHZNXHUCAGE-UHFFFAOYSA-N
2-(1,2,4-triazol-1-ylmethyl)aniline
6482003
97%
127988-21-0
100.0
Amber Glass Bottle
MFCD09025845
2-1h-1,2,4-triazol-1-ylmethyl aniline, 2-1,2,4-triazol-1-yl methyl aniline, 2-1,2,4-triazol-1-ylmethyl aniline, 2-1h-1,2,4-triazol-1-ylmethyl benzeneamine, benzenamine,2-1h-1,2,4-triazol-1-ylmethyl, acmc-20msof, 2-1,2,4-triazolylmethyl phenylamine
C1=CC=C(C(=C1)CN2C=NC=N2)N
174.207
174.21

Chemical Identifiers

127988-21-0
174.207
CBGQHZNXHUCAGE-UHFFFAOYSA-N
6482003
C1=CC=C(C(=C1)CN2C=NC=N2)N
C9H10N4
MFCD09025845
2-1h-1,2,4-triazol-1-ylmethyl aniline, 2-1,2,4-triazol-1-yl methyl aniline, 2-1,2,4-triazol-1-ylmethyl aniline, 2-1h-1,2,4-triazol-1-ylmethyl benzeneamine, benzenamine,2-1h-1,2,4-triazol-1-ylmethyl, acmc-20msof, 2-1,2,4-triazolylmethyl phenylamine
2-(1,2,4-triazol-1-ylmethyl)aniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions