Learn More
2-[1-(tert-Butoxycarbonyl)piperid-4-yl]-1,3-thiazole-4-carboxylic acid, ≥95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC31001DA
| Quantity | 1g |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 668484-45-5 | |
| 313.372 | |
| FMSFMUBALWPCQM-UHFFFAOYSA-N | |
| 59496772 | |
| CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CS2)C(=O)O |
| C13H19N3O4S | |
| MFCD06658980 | |
| 2-4-tert-butoxycarbonyl piperazin-1-yl thiazole-4-carboxylic acid, 2-4-tert-butoxycarbonyl piperazin-1-yl-1,3-thiazole-4-carboxylic acid | |
| 2-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]-1,3-thiazole-4-carboxylic acid |
Specifications
| 2-[1-(Tert-butoxycarbonyl)-4-piperidinyl]-1, 3-thiazole-4-carboxylic acid | |
| 94.5 | |
| 95% | |
| C13H19N3O4S | |
| MFCD06658980 | |
| FMSFMUBALWPCQM-UHFFFAOYSA-N | |
| 2-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]-1,3-thiazole-4-carboxylic acid | |
| 59496772 | |
| 95% |
| 668484-45-5 | |
| 100.0 | |
| Amber Glass Bottle | |
| 1g | |
| 2-4-tert-butoxycarbonyl piperazin-1-yl thiazole-4-carboxylic acid, 2-4-tert-butoxycarbonyl piperazin-1-yl-1,3-thiazole-4-carboxylic acid | |
| CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CS2)C(=O)O | |
| 313.372 | |
| 312.39 |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.