missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Pyrrolyl)benzoic acid, 99%

Catalog No. AAL0606903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0606903 1 g
AAL0606906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0606903 Supplier Thermo Scientific Chemicals Supplier No. L0606903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10333-68-3 | C11H9NO2 | 187.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10333-68-3
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD00051645
InChI Key GNWTWXOZRSBCOZ-UHFFFAOYSA-N
Synonym 2-1h-pyrrol-1-yl benzoic acid, 2-1-pyrrolyl benzoic acid, 1-2-carboxyphenyl pyrrole, 2-pyrrol-1-yl benzoic acid, 2-pyrrol-1-yl-benzoic acid, 2-pyrrolylbenzoic acid, maybridge1_002677, 1-o-carboxyphenyl pyrrole, n-2-carboxyphenyl pyrrole, 1h-pyrrol-1-ylbenzoic acid
PubChem CID 728521
IUPAC Name 2-pyrrol-1-ylbenzoic acid
SMILES OC(=O)C1=CC=CC=C1N1C=CC=C1

Specifications

Melting Point 102°C to 106°C
Quantity 1 g
Beilstein 1343920
Formula Weight 187.2
Percent Purity 99%
Chemical Name or Material 2-(1-Pyrrolyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only