missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperidinyl)benzonitrile, 97%

Catalog No. p-7081705
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1178403 1 g
AAA1178406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1178403 Supplier Thermo Scientific Chemicals Supplier No. A1178403
Only null left

CAS: 72752-52-4 | C12H14N2 | 186.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72752-52-4
Molecular Formula C12H14N2
Molecular Weight (g/mol) 186.258
MDL Number MFCD00049221
InChI Key MEBVSLLKZSAIGK-UHFFFAOYSA-N
Synonym 2-piperidinobenzonitrile, 2-piperidin-1-yl benzonitrile, 2-1-piperidinyl benzonitrile, gnf-pf-3161, benzonitrile, 2-1-piperidinyl, 2-piperidylbenzenecarbonitrile, 2-piperidino benzonitrile, maybridge1_000930, acmc-1bfz1, 2-1-piperidyl benzonitrile
PubChem CID 2774355
IUPAC Name 2-piperidin-1-ylbenzonitrile
SMILES C1CCN(CC1)C2=CC=CC=C2C#N

Specifications

Melting Point 41°C to 45°C
Boiling Point 122°C to 125°C (1 mmHg)
Quantity 1 g
UN Number UN3077
Beilstein 1243909
Formula Weight 186.26
Percent Purity 97%
Chemical Name or Material 2- (1-Piperidinyl)benzonitrile
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 427-330-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only