Learn More
2-(1-Piperazinyl)pyrimidine, 99%
CAS: 20980-22-7 | C8H12N4 | 164.212 g/mol
$60.74 - $434.26
Chemical Identifiers
| CAS | 20980-22-7 |
|---|---|
| Molecular Formula | C8H12N4 |
| Molecular Weight (g/mol) | 164.212 |
| MDL Number | MFCD00040742 |
| InChI Key | MRBFGEHILMYPTF-UHFFFAOYSA-N |
| Synonym | 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine |
| PubChem CID | 88747 |
| IUPAC Name | 2-piperazin-1-ylpyrimidine |
| SMILES | C1CN(CCN1)C2=NC=CC=N2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1588406
|
Thermo Scientific Chemicals
L1588406 |
5 g |
Each for $60.74
|
|
|||||
|
AAL1588414
|
Thermo Scientific Chemicals
L1588414 |
25 g |
Each for $146.00
|
|
|||||
|
AAL1588422
|
Thermo Scientific Chemicals
L1588422 |
100 g |
Each for $434.26
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 20980-22-7 | |
| 164.212 | |
| MRBFGEHILMYPTF-UHFFFAOYSA-N | |
| 88747 | |
| C1CN(CCN1)C2=NC=CC=N2 |
| C8H12N4 | |
| MFCD00040742 | |
| 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine | |
| 2-piperazin-1-ylpyrimidine |
Specifications
| 20980-22-7 | |
| 118°C to 120°C (2 mmHg) | |
| MFCD00040742 | |
| UN3259 | |
| Air sensitive; Hygroscopic | |
| MRBFGEHILMYPTF-UHFFFAOYSA-N | |
| 2-piperazin-1-ylpyrimidine | |
| 88747 | |
| 99% |
| 31°C to 35°C | |
| C8H12N4 | |
| 5 g | |
| 151178 | |
| 2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine | |
| C1CN(CCN1)C2=NC=CC=N2 | |
| 164.212 | |
| 164.21 | |
| 2-(1-Piperazinyl)pyrimidine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber : 244-135-5
RTECSNumber : UV9702000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only