missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthylmethyl)-2-imidazoline hydrochloride, 99%

Catalog No. p-7027421
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2416114 25 g
AAB2416122 100 g
AAB2416136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2416114 Supplier Thermo Scientific Chemicals Supplier No. B2416114
Only null left

CAS: 550-99-2 | C14H15ClN2 | 246.738 g/mol

Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphazoline hydrochloride produces ionotropic effects via α-adrenoceptor stimulation. It is an activator of AR and Nischarin. It is used as a nasal decongestant and ocular vasoconstrictor.

Solubility
Soluble in water 170 g/L, alcohol, DMSO, ethanol.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 550-99-2
Molecular Formula C14H15ClN2
Molecular Weight (g/mol) 246.738
MDL Number MFCD00012554
InChI Key DJDFFEBSKJCGHC-UHFFFAOYSA-N
Synonym naphazoline hydrochloride, albalon, naphazoline hcl, rhinantin, rhinoperd, stricylon, naphcon, niazol, rinofug, vasocon
PubChem CID 11079
ChEBI CHEBI:7470
IUPAC Name 2-(naphthalen-1-ylmethyl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES C1CN=C(N1)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

Melting Point 257°C to 261°C
Quantity 25 g
UN Number UN2811
Beilstein 3716843
Merck Index 14,6368
Solubility Information Soluble in water 170g/L,alcohol,DMSO,ethanol.
Formula Weight 246.74
Percent Purity 99%
Chemical Name or Material 2-(1-Naphthylmethyl)-2-imidazoline hydrochloride
Hazard Category H300
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 208-989-2
RTECSNumber NJ4375000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only