missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)-5-phenyloxazole, laser grade and suitable for scintillation spectrometry, 99+%

Catalog No. p-7023283
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1667903 1 g
AAA1667906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1667903 Supplier Thermo Scientific Chemicals Supplier No. A1667903
Only null left

CAS: 846-63-9 | C19H13NO | 271.319 g/mol

Starting with the organic fluor 2-(1-naphthyl)-5-phenyloxazole (αNPO), the vNPO and allylNPO monomers were synthesized using Stille coupling followed by purification. Used as a laser dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Starting with the organic fluor 2-(1-naphthyl)-5-phenyloxazole (αNPO), the vNPO and allylNPO monomers were synthesized using Stille coupling followed by purification. Used as a laser dye.

Solubility
Sparingly Soluble (4.1E-4 g/L) (25°C). Soluble most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 846-63-9
Molecular Formula C19H13NO
Molecular Weight (g/mol) 271.319
MDL Number MFCD00019714
InChI Key WWVFJJKBBZXWFV-UHFFFAOYSA-N
Synonym 2-1-naphthyl-5-phenyloxazole, alpha-npo, oxazole, 2-1-naphthalenyl-5-phenyl, 2-1-naphthyl-5-phenyl-1,3-oxazole, 2-naphthalen-1-yl-5-phenyloxazole, oxazole, 2-1-naphthyl-5-phenyl, .alpha.-npo, anpo, 2-alpha-naphthyl-5-phenyloxazole, 2-naphthalen-1-yl-5-phenyl-1,3-oxazole
PubChem CID 70058
IUPAC Name 2-naphthalen-1-yl-5-phenyl-1,3-oxazole
SMILES C1=CC=C(C=C1)C2=CN=C(O2)C3=CC=CC4=CC=CC=C43

Specifications

Melting Point 104°C to 105°C
Quantity 1 g
Beilstein 210666
Solubility Information Sparingly Soluble (4.1E-4g/L) (25°C). Soluble most organic solvents.
Formula Weight 271.32
Percent Purity ≥99%
Grade Suitable for scintillation spectrometry
Chemical Name or Material 2-(1-Naphthyl)-5-phenyloxazole
EINECSNumber 212-689-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only