missing translation for 'onlineSavingsMsg'
Learn More

{2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]phenyl}methanol, 97%, Thermo Scientific™

Catalog No. CC62109CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC62109CB 250mg Amber Glass Bottle
CC62109DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC62109CB Supplier Thermo Scientific Chemicals Supplier No. CC62109CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 892502-29-3
Molecular Formula C12H11F3N2O
Molecular Weight (g/mol) 256.228
MDL Number MFCD09817511
InChI Key HLMRFDCLSUXVQC-UHFFFAOYSA-N
Synonym 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl phenyl methanol, 2-2-methyl-5-trifluoromethyl pyrazol-3-yl phenyl methanol, 2-1-methyl-3-trifluoromethyl pyrazol-5-yl phenyl methan-1-ol, 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzyl alcohol, benzenemethanol,2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl
PubChem CID 24229659
IUPAC Name [2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]phenyl]methanol
SMILES CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 256.23
Percent Purity 97%
Chemical Name or Material {2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]phenyl}methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.