missing translation for 'onlineSavingsMsg'
Learn More

2-[1-methyl-3-(trifluoromethyl)-1h-pyrazol-5-yl]benzoic acid, 97%, Thermo Scientific™

Catalog No. p-3586647
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC62101CB 250mg Amber Glass Bottle
CC62101DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC62101CB Supplier Thermo Scientific Chemicals Supplier No. CC62101CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 910037-16-0
Molecular Formula C12H9F3N2O2
Molecular Weight (g/mol) 270.211
MDL Number MFCD09879926
InChI Key ADEAPHCAMGDLFU-UHFFFAOYSA-N
Synonym 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzoic acid, 2-2-methyl-5-trifluoromethyl pyrazol-3-yl benzoic acid
PubChem CID 24229655
IUPAC Name 2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]benzoic acid
SMILES CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 270.21
Percent Purity 97%
Chemical Name or Material 2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.