Learn More
2-(1-Methoxy)propyl acetate, 99%
CAS: 108-65-6 | C6H12O3 | 132.16 g/mol
$156.44 - $156.44
Chemical Identifiers
| CAS | 108-65-6 |
|---|---|
| Molecular Formula | C6H12O3 |
| Molecular Weight (g/mol) | 132.16 |
| MDL Number | MFCD00038500 |
| InChI Key | LLHKCFNBLRBOGN-UHFFFAOYSA-N |
| Synonym | 1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester |
| PubChem CID | 7946 |
| IUPAC Name | 1-methoxypropan-2-yl acetate |
| SMILES | CC(COC)OC(=O)C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC413900010
|
Thermo Scientific Chemicals
413900010 |
1 kg | Glass bottle |
Each for $156.44
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 108-65-6 | |
| 132.16 | |
| LLHKCFNBLRBOGN-UHFFFAOYSA-N | |
| 7946 | |
| CC(COC)OC(=O)C |
| C6H12O3 | |
| MFCD00038500 | |
| 1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester | |
| 1-methoxypropan-2-yl acetate |
Specifications
| 108-65-6 | |
| 100.0 | |
| Colorless | |
| 146.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4010 to 1.4030 | |
| MFCD00038500 | |
| 1993 | |
| 1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester | |
| LLHKCFNBLRBOGN-UHFFFAOYSA-N | |
| 1-methoxypropan-2-yl acetate | |
| 7946 | |
| 99% | |
| 2-(1-Methoxy)propyl acetate |
| 98.5 | |
| -87.0°C | |
| 0.9600g/mL | |
| 42°C | |
| 98.5% min. (GC) | |
| C6H12O3 | |
| CH3OCH2CH(CH3)OC(O)CH3 | |
| 1 kg | |
| 0.96 | |
| Solubility in water: 19.8g/L (25°C) | |
| CC(COC)OC(=O)C | |
| 132.16 | |
| 132.16 | |
| Liquid |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
GHS Signal Word: Warning
EINECSNumber : 203-603-9
RUO – Research Use Only