missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Methoxy)propyl acetate, 99%

Catalog No. AC413900010
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC413900010 1 kg Glass bottle
AC413900050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC413900010 Supplier Thermo Scientific Chemicals Supplier No. 413900010

CAS: 108-65-6 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-65-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00038500
InChI Key LLHKCFNBLRBOGN-UHFFFAOYSA-N
Synonym 1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl ester
PubChem CID 7946
IUPAC Name 1-methoxypropan-2-yl acetate
SMILES CC(COC)OC(=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -87.0°C
Color Colorless
Density 0.9600g/mL
Boiling Point 146.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4010 to 1.4030
Linear Formula CH3OCH2CH(CH3)OC(O)CH3
Quantity 1 kg
UN Number 1993
Specific Gravity 0.96
Solubility Information Solubility in water: 19.8g/L (25°C)
Formula Weight 132.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(1-Methoxy)propyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge
EINECSNumber 203-603-9

RUO – Research Use Only