missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Cyclohexenyl)ethylamine 98.0+%, TCI America™

Catalog No. C1157500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1157500ML 500 mL
C115725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1157500ML Supplier TCI America Supplier No. C1157500ML

Chemical Identifiers

CAS 3399-73-3
Molecular Formula C8H15N
Molecular Weight (g/mol) 125.215
MDL Number MFCD00012058
InChI Key IUDMXOOVKMKODN-UHFFFAOYSA-N
Synonym 2-1-cyclohexenyl ethylamine, 1-cyclohexene-1-ethanamine, 2-cyclohex-1-en-1-yl ethanamine, cyclohexenylethylamine, 1-cyclohexen-1-ylethylamine, 2-1-cyclohexen-1-yl ethylamine, cyclohex-1-ene-1-ethylamine, 2-cyclohex-1-en-1-yl ethan-1-amine, 2-cyclohexen-1-yl ethanamine, .beta.-1-cyclohexenyl ethylamine
PubChem CID 76938
IUPAC Name 2-(cyclohexen-1-yl)ethanamine
SMILES C1CCC(=CC1)CCN

Specifications

Color Yellow
Boiling Point 84°C
Quantity 500 mL
UN Number 2733
Formula Weight 125.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(1-Cyclohexenyl)ethylamine