missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Cyclohexenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™

Catalog No. C27071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27071G Supplier TCI America Supplier No. C27071G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141091-37-4
Molecular Formula C12H21BO2
Molecular Weight (g/mol) 208.11
MDL Number MFCD05663845
InChI Key QNZFUMVTUFOLRT-UHFFFAOYSA-N
Synonym 2-cyclohex-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexen-yl-boronic acid pinacol ester, cyclohexene-1-boronic acid pinacol ester, 1-cyclohexen-1-yl-boronic acid pinacol ester, 2-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid, 2-1-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, cyclohexen-1-ylboronic acid, pinacol ester, 2-1-cyclohexen-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid pinacol ester
PubChem CID 10932675
IUPAC Name 2-(cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCCCC1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 208.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(1-Cyclohexenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
TSCA No
Recommended Storage Refrigerator